Orders LinkContactUsLinkComponent
Accessories for HOBO State Data Logger
Accessories for HOBO State Data Logger
Split-core AC current transformer sensor, 10 - 100 AMP, ±1% linearity
Catalog # 470324-686
Supplier #:  T-MAG-SCT-100
Accessories for HOBO State Data Logger
Catalog # 470324-686
Accessories for HOBO State Data Logger
Accessories for HOBO State Data Logger
Split-core AC current transformer sensor, 10 - 100 AMP, ±1% linearity
Price
$160.29
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Expands the measurement capabilities of a HOBO State Data Logger by enabling reliable monitoring of electrical current and equipment operation without interrupting existing systems.
  • Split-core design allows installation without disconnecting power
    Measures AC current for equipment and energy monitoring applications
    Produces proportional output for accurate state and load detection
    Compatible with HOBO state data logger accessories and interface modules
    Supports monitoring of electrical equipment operation and usage patterns
The accessories for the HOBO state data logger are designed to expand the range of applications that can be monitored using a single logging platform. By adding compatible sensors and interface components, users can capture electrical current and operational state data that are not directly measurable by the logger alone. These accessories support monitoring of equipment run times, load changes, and on/off events, making them useful for applications such as energy assessments, equipment diagnostics, and operational analysis.
One commonly used accessory is a split-core AC current transformer sensor, which allows electrical current to be measured safely and efficiently without disconnecting power. The sensor clamps around an existing conductor and produces a proportional output that can be interpreted by compatible HOBO interface modules. This design simplifies installation in retrofit or active systems and minimizes disruption to ongoing operations.
When used together, these accessories enable more comprehensive data collection for facilities management, research, and industrial monitoring projects. They help users build a clearer picture of system performance and usage patterns while maintaining the flexibility and reliability expected from the HOBO monitoring ecosystem.
Specifications
  • Description:
    Split-core AC current transformer sensor, 10 - 100 AMP
  • Accuracy:
    ±1% linearity