Orders LinkContactUsLinkComponent
Thumb Dressing Forceps, Electron Microscopy Sciences
Thumb Dressing Forceps, Electron Microscopy Sciences
Fine grip forceps, 140 mm, Serrated tips
Catalog # 76375-982
Supplier #:  77937-23
Thumb Dressing Forceps, Electron Microscopy Sciences
Catalog # 76375-982
Thumb Dressing Forceps, Electron Microscopy Sciences
Thumb Dressing Forceps, Electron Microscopy Sciences
Fine grip forceps, 140 mm, Serrated tips
Price
$125.78
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These conventional forceps are very popular and are available in a variety of length, tip, and handle styles.
  • Made of premium grade, hardened stainless steel
  • Feature a protective coating for extra durability and protection from rust and corrosion
  • Satin-matte finish reduces eye-fatiguing glare
Specifications
  • Description:
    Fine grip forceps
  • Tip configuration:
    Serrated tips
  • Overall length:
    140 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)