$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
SEASPRAY DC NEBULIZER 2ML/MIN W/TUBING
103788-766
:
Glass Expansion Inc
SEASPRAY DC NEBULIZER 2ML/MIN W/TUBING
103788-766
:
Glass Expansion Inc
:
A41-1-USS04
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International