Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SINK BASE UNIT LEFT DOOR 22X18X29-1/8IN
Catalog # 76360-728
Supplier:
Eagle Group
Expand image
SINK BASE UNIT LEFT DOOR 22X18X29-1/8IN
Catalog # 76360-728
Supplier:
Eagle Group
Supplier Number:
SSC-SSBL-18
Related Information
Documentation
Datasheet
TDS Sitting Height Base Cabinets Freestanding Sink Bases and Angled Sink Bases.pdf
Documentation
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH