Order Entry
Canada
Orders LinkContactUsLinkComponent
Treston® Concept Lab Bench Accessories

Treston® Concept Lab Bench Accessories

Shelf lip with 3" edge (seismic), light gray, 36" W, Concept and cornerstone
Catalog # 76365-628
Supplier:   Treston
Supplier #:  890704-49
Treston® Concept Lab Bench Accessories
Catalog # 76365-628
Supplier:  Treston
Treston® Concept Lab Bench Accessories
Treston® Concept Lab Bench Accessories

Shelf lip with 3" edge (seismic), light gray, 36" W, Concept and cornerstone

Price
$58.19
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Interchangeable accessories across all Treston bench frames.

  • Attached with the aid of upright tubes or a complete upright tube module
  • Compatible on all Treston frames

Ordering information: Installation is required. Assembly videos available.

Specifications
  • Description:
    Shelf lip with 3" edge (seismic), light gray, 36" W
  • For use with:
    Concept and cornerstone