Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Polytron® Generator, PT-DA/2WEC-E192, Kinematica

Polytron® 20 mm generator, PT-DA 20/2WEC-E192, 10 - 2500 ml, 20×192 mm, PT 2500 E
  76360-442
 :   Kinematica
 :  11030258
Polytron® Generator, PT-DA/2WEC-E192, Kinematica
  76360-442
Polytron® Generator, PT-DA/2WEC-E192, Kinematica
Polytron® Generator, PT-DA/2WEC-E192, Kinematica

Polytron® 20 mm generator, PT-DA 20/2WEC-E192, 10 - 2500 ml, 20×192 mm, PT 2500 E

 
$5,599.47
   
Each
 

W-design for fibrous/stringy materials. Special design to prevent the teeth clogging. Dispersing generators are made from high-alloy stainless steel 316L, electropolished, Ra≤1.6 μm as standard.

  • Dispersing fibrous, stringy or solid samples (e.g., pieces of meat)
  • Standard dispersing, emulsifying and suspending tasks
 
  • Description:
    Polytron® 20 mm generator, PT-DA 20/2WEC-E192
  • Diameter ×L:
    20×192 mm
  • For use with:
    PT 2500 E
  • Sample volume:
    10 - 2500 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.