Order Entry
Canada
ContactUsLinkComponent
CELL SCREEN SS 400 MESH 38U 37 MM CS5
Catalog # 76356-940
Supplier:  Bellco Glass
undefined
CELL SCREEN SS 400 MESH 38U 37 MM CS5
Catalog # 76356-940
Supplier:  Bellco Glass
Supplier Number:  1985-00400
Frequently Bought Together