Order Entry
Canada
ContactUsLinkComponent
IOB W/MICRO-CARRIER FLASK 100MM 45MM 36L
Catalog # 76356-632
Supplier:  Bellco Glass
undefined
IOB W/MICRO-CARRIER FLASK 100MM 45MM 36L
Catalog # 76356-632
Supplier:  Bellco Glass
Supplier Number:  1965-61360
Frequently Bought Together