Order Entry
Canada
ContactUsLinkComponent
IOB W/MICRO-CARRIER FLASK 100MM 1L
Catalog # 76356-620
Supplier:  Bellco Glass
undefined
IOB W/MICRO-CARRIER FLASK 100MM 1L
Catalog # 76356-620
Supplier:  Bellco Glass
Supplier Number:  1965-61010
Frequently Bought Together