Passer à En-tête
Passer à Contenu principal
Passer à Pied de page
Avancé
Avancé
SDS
Certificats
Données du fabricant
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GLOVES RIGHT STEELCORE 2 7GAUGE SS SMALL
Catalog # 76349-546
Supplier:
MCR Safety
Expand image
GLOVES RIGHT STEELCORE 2 7GAUGE SS SMALL
Catalog # 76349-546
Supplier:
MCR Safety
Supplier Number:
9381SRH
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International