Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RIDER COAT BLACK 0.35MM 60IN PVC/PES 3XL
Catalog # 76342-040
Supplier:
MCR Safety
Expand image
RIDER COAT BLACK 0.35MM 60IN PVC/PES 3XL
Catalog # 76342-040
Supplier:
MCR Safety
Supplier Number:
FR267CX3
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International