$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
SAFETY VEST CL2 BREAKAWAY 2PKT LF 2IN X2
76340-158
:
MCR Safety
SAFETY VEST CL2 BREAKAWAY 2PKT LF 2IN X2
76340-158
:
MCR Safety
:
CL2MLPFRX2
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International