$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
SAFETY VEST CL2 TEAR-AWAY X-BACK 2IN X2
Catalog # 76340-122
Supplier:
MCR Safety
Expand image
SAFETY VEST CL2 TEAR-AWAY X-BACK 2IN X2
Catalog # 76340-122
Supplier:
MCR Safety
Supplier Number:
XCL2MLX2
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International