Orders LinkContactUsLinkComponent
Ward's® Plastic Specimen Trays
WARD’S Plastic Specimen Trays, 36-Compartment Tray
  470320-674
 :  B19180/25-SP
Ward's® Plastic Specimen Trays
  470320-674
 :  
Ward's® Plastic Specimen Trays
Ward's® Plastic Specimen Trays
WARD’S Plastic Specimen Trays, 36-Compartment Tray
Keep Your Collections Organized
Use our vacuum-formed compartmented trays alone to keep small items separate, or in our collection boxes for permanent dustfree storage of your samples. The white nonglare finish will not yellow with age.
12-Compartment Tray
Tray size: 6 3/4"L x 6 3/4"W x 5/8"H; cavity size: 2"L x 1 1/2"W.
16-Compartment Tray
Tray size: 7"L x 7"W x 5/8"H; cavity size: 1 1/2"L x 1 1/2"W.
24-Compartment Tray
Tray size: 12 1/2"L x 9 1/4"W x 5/8"H; cavity size: 2 1/4"L x 1 7/8"W.
36-Compartment Tray
Tray size: 12 1/2"L x 9 1/4"W x 5/8"H; cavity size: 2"L x 1 1/2"W.
45-Compartment Tray
Tray size: 16"L x 12 1/2"W x 5/8"H; cavity size: 2 1/2"L x 1 3/4"W.
50-Compartment Tray
Tray size: 15 3/4"L x 12 1/2"W x 3/4"H; cavity size: 2 1/4"L x 1 3/8"W.
  • Description:
    WARD’S Plastic Specimen Trays, 36-Compartment Tray