Orders LinkContactUsLinkComponent
Embedding Blocks
Embedding Blocks
Embedding blocks, 1×1׳/₄"
Catalog # 470320-808
Supplier #:  EB0004
Embedding Blocks
Catalog # 470320-808
Embedding Blocks
Embedding Blocks
Embedding blocks, 1×1׳/₄"
Price
$150.75
Log in for availability
Pack of 1
Minimum order quantity is 1
Product Details & Documents
Embedding blocks for biological specimens.
  • Wide range of sizes
  • Made of phenolic resin
Used for celloidin or paraffin sections, these embedding blocks are made of phenolic resin, which can be marked with a glass marking pencil.  Top surface has 1/32" deep grid to provide a firm hold for embedding specimen.  Blocks are ¾" high. Sold in packs of 10.
Specifications
  • Description:
    Embedding blocks
  • Dimensions:
    1×1׳/₄"