Embedding Blocks
Embedding blocks, ⁵/₈×⁵/₈׳/₄"
Catalog # 470320-804
Supplier: United Scientific Supplies
: EB0002
Jump to:
Product Details & Documents
Embedding blocks for biological specimens.
- Wide range of sizes
- Made of phenolic resin
Used for celloidin or paraffin sections, these embedding blocks are made of phenolic resin, which can be marked with a glass marking pencil. Top surface has 1/32" deep grid to provide a firm hold for embedding specimen. Blocks are ¾" high. Sold in packs of 10.
Specifications
- Description:Embedding blocks
- Dimensions:⁵/₈×⁵/₈׳/₄"