Orders LinkContactUsLinkComponent
Embedding Blocks
Embedding Blocks
Embedding blocks, ¹/₂×¹/₂׳/₄"
Catalog # 470320-802
Supplier #:  EB0001
Embedding Blocks
Catalog # 470320-802
Embedding Blocks
Embedding Blocks
Embedding blocks, ¹/₂×¹/₂׳/₄"
Price
$61.95
Log in for availability
Pack of 1
Minimum order quantity is 1
Product Details & Documents
Embedding blocks for biological specimens.
  • Wide range of sizes
  • Made of phenolic resin
Used for celloidin or paraffin sections, these embedding blocks are made of phenolic resin, which can be marked with a glass marking pencil.  Top surface has 1/32" deep grid to provide a firm hold for embedding specimen.  Blocks are ¾" high. Sold in packs of 10.
Specifications
  • Description:
    Embedding blocks
  • Dimensions:
    ¹/₂×¹/₂׳/₄"