Order Entry
Canada
Orders LinkContactUsLinkComponent
Ammonia solution 2%

Ammonia solution 2%

Poly bottle, 4 L
Catalog # RCR0631000-4A
Supplier:   Ricca Chemical
Supplier #:  R0631000-4A
Ammonia solution 2%
Catalog # RCR0631000-4A
Supplier:  Ricca Chemical
CAS Number:  1336-21-6
Ammonia solution 2%
Ammonia solution 2%

Poly bottle, 4 L

CAS Number:  1336-21-6
Price
$255.51
Log in for availability
Each (4l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:100 °C
  • Melting Pt:0.0 °C
  • Density:0.9978 g/cm³
  • Storage Temperature:Ambient

Documents

Specifications

Specification Test Results

  • Appearance:
    Colorless liquid
  • Assay (vs. Sulfuric Acid/Methyl Red):
    1.90 - 2.10 % (v/v)

Specifications

  • Pack type:
    Poly bottle
  • Size:
    4 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.