Order Entry
Canada
Orders LinkContactUsLinkComponent
pH Buffer Capsules, Oakton® Instruments

pH Buffer Capsules, Oakton® Instruments

pH capsules, 100 ml, ±0.02, 25 °C, 10 capsules/vial, 10.00
Catalog # 76325-410
Supplier:   Antyila Scientific
Supplier #:  35850-16
pH Buffer Capsules, Oakton® Instruments
Catalog # 76325-410
Supplier:  Antyila Scientific
CAS Number:  
pH Buffer Capsules, Oakton® Instruments
pH Buffer Capsules, Oakton® Instruments

pH capsules, 100 ml, ±0.02, 25 °C, 10 capsules/vial, 10.00

CAS Number:  
Price
$41.55
New Customer Promotional Price
Log in for availability
Pack of 10
Product Details & Documents

Fresh pH calibration solution anytime and anywhere.

  • One capsule to 100 ml of deionized or distilled water
  • Dry and compact—easily take them anywhere
  • Unused capsules have a five-year shelf life
  • Each vial contains 10 capsules

Buffer salts inside capsules require 100 mL of deionized or distilled water to produce a clear pH calibration solution accurate to ±0.02 pH (at 25°C) with traceability to NIST standards. Capsules are easier to store and transport than prepared aqueous buffer solutions. Capsules are color-coded for easy identification.

Once dissolved, store buffer solution in an airtight container at room temperature for up to three weeks.

Specifications

Specifications

  • Description:
    pH capsules
  • Size:
    10 capsules/vial
  • Reference temperature:
    25 °C
  • pH:
    10.00
  • Final volume:
    100 ml
  • pH accuracy:
    ±0.02

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.