Order Entry
Canada
Orders LinkContactUsLinkComponent
 

pH Buffer Capsules, Oakton® Instruments

pH capsules, 100 ml, ±0.02, 25 °C, 10 capsules/vial, 4.00
Catalog # 76325-406
Supplier:   Antyila Scientific
 :  35850-12
pH Buffer Capsules, Oakton® Instruments
Catalog # 76325-406
Supplier:  Antyila Scientific
CAS Number:  
pH Buffer Capsules, Oakton® Instruments
pH Buffer Capsules, Oakton® Instruments

pH capsules, 100 ml, ±0.02, 25 °C, 10 capsules/vial, 4.00

CAS Number:  
Price
$41.55
New Customer Promotional Price
Log in for availability
Pack of 10
Product Details & Documents

Fresh pH calibration solution anytime and anywhere.

  • One capsule to 100 ml of deionized or distilled water
  • Dry and compact—easily take them anywhere
  • Unused capsules have a five-year shelf life
  • Each vial contains 10 capsules

Buffer salts inside capsules require 100 mL of deionized or distilled water to produce a clear pH calibration solution accurate to ±0.02 pH (at 25°C) with traceability to NIST standards. Capsules are easier to store and transport than prepared aqueous buffer solutions. Capsules are color-coded for easy identification.

Once dissolved, store buffer solution in an airtight container at room temperature for up to three weeks.

Specifications

Specifications

  • Description:
    pH capsules
  • Size:
    10 capsules/vial
  • Reference temperature:
    25 °C
  • pH:
    4.00
  • Final volume:
    100 ml
  • pH accuracy:
    ±0.02

Specifications