Order Entry
Canada
Orders LinkContactUsLinkComponent
Dissecting Forceps

Dissecting Forceps

Fine point forceps, Stainless steel, 115 mm (4.5"), Straight
Catalog # 470315-182
Supplier #:  15C
Dissecting Forceps
Catalog # 470315-182
Dissecting Forceps
Dissecting Forceps

Fine point forceps, Stainless steel, 115 mm (4.5"), Straight

Price
8,20 $
Log in for availability
Each Box of 100
Product Details & Documents

Quality fine point forceps.

  • Straight or bent tip
  • Stainless steel construction
  • Textured grip

Dissecting forceps feature straight fine points. Made with high quality stainless steel. Fine points are great for delicate dissection purposes and can be used under the microscope.

Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.

Specifications
  • Description:
    Fine point forceps
  • Length:
    115 mm (4.5")
  • Material:
    Stainless steel
  • Tip style:
    Straight
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.