Order Entry
Canada
Orders LinkContactUsLinkComponent
Iris Scissors

Iris Scissors

Iris Scissors, 115 mm (4.5"), Stainless steel, Curved
Catalog # 470315-164
Supplier #:  10
Iris Scissors
Catalog # 470315-164
Iris Scissors
Iris Scissors

Iris Scissors, 115 mm (4.5"), Stainless steel, Curved

Price
$9.90
Log in for availability
Each Box of 25
Product Details & Documents

For fine dissections.

  • Used for very fine dissections, and working in small areas

The blades are very narrow allowing for a very clean cut, and a short blade length gives greater control. These iris scissors have excellent rust resistance. The scissors marked TC, are tungsten carbon tipped for greater edge retention and strength.

Ordering information: Available in curved or straight configuration.

Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.

Specifications
  • Description:
    Iris Scissors
  • Length:
    115 mm (4.5")
  • Material:
    Stainless steel
  • Curvature:
    Curved
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.