VWR® Gel-Loading Pipette Tips
Round MBP Style, Ø 0.57 mm, 200 µl, 68 mm, Bulk
Supplier: VWR International
Jump to:
Product Details & Documents
VWR® Gel-Loading Pipette Tips faciliate precision sample loading into gels.
- Standard 96 tip format is multi-channel friendly
- Certified free of RNase, DNase, and human DNA (lot-by-lot certificates available)
- Sterile product is also pyrogen-free
Universal fit to accommodate most pipettes except Rainin LTS and Thermo Clip-Tip.
Ordering information: Choose from sterile or nonsterile formats, racked or bulk packaging, slim or MBP body styles.
Documents
Certificates
Search by Lot NumberSpecifications
- Volume:200 µl
- Length:68 mm
- Packed:Bulk
- Tip style:Round MBP Style, Ø 0.57 mm



