Axygen® Multirack Bulk Tips, Corning
Standard Length, 1 - 200 µl, Bulk: 1000 Tips/Bag, 10 Bags/Case
Supplier: Corning
Supplier #: MR-200
Jump to:
Product Details & Documents
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an inter-changeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
- Detectable DNase/RNase/Human DNA/PCR inhibition free
- Nonpyrogenic
- Manufactured in an ISO certified facility in the USA
- Fully autoclavable
Axygen® MultiRack bulk pipette tips are compatible with MultiRack racks and packaged in resealable polyethylene bags, non sterile.
Ordering information: Axygen® MultiRack pipette tips are also available in hinged rack, filter tips, reload, and tipstation configurations.
Specifications
- Volume:1 - 200 µl
- Packed:Bulk: 1000 Tips/Bag, 10 Bags/Case
- Tip style:Standard Length
Accessories
Product Information
- Catalog #For Use WithAvailabilityPrice
- MultiRack Empty Rack, 5/Pack, 1 Pack/CaseMultiRack Empty Rack, 5/Pack, 1 Pack/Case76320-74410 µl extended length, 200 µl, 200 µl NX, 300 µl tipsLog in for availabilityCase of 5 - $133.50
Product Information
- Catalog #For Use WithAvailabilityPrice
- MultiRack Empty Rack, 5/Pack, 1 Pack/CaseMultiRack Empty Rack, 5/Pack, 1 Pack/Case76320-746200 µl extended length, 300 µl extended length, and 1000 µl extended length tipsLog in for availabilityCase of 5 - $180.00