Camera Adapters
C-mount adapter without lens, 1.00×, —
Catalog # 470314-900
Supplier: Motic
Supplier #: 1101001901771
Jump to:
Product Details & Documents
C-mount camera adapter allows Moticams to connect with Motic trinocular microscopes.
Specifications
- Description:C-mount adapter without lens, 1.00×
- For use with:—