Orders LinkContactUsLinkComponent
Biox Micro PettePlus Pipettes
Biox Micro PettePlus Pipettes
MicroPette Plus Pipette, 100 - 1000 µl, 5 µl
Catalog # 76319-900
Supplier:   TWD Scientific
Supplier #:  BX01-2110
Biox Micro PettePlus Pipettes
Catalog # 76319-900
Supplier:  TWD Scientific
Biox Micro PettePlus Pipettes
Biox Micro PettePlus Pipettes
MicroPette Plus Pipette, 100 - 1000 µl, 5 µl
Price
$228.57
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
MicroPette Plus mechanical pipettes are the ideal adjustable volume pipettes for all of your basic pipetting needs.
  • Ergonomic design for comfortable extended use
  • Easy-to-read display
  • Convenient one-handed operation
  • Easy to calibrate and maintain with included tool
  • Individual piston and tip cone assemblies allow for easy repair and maintenance
  • Fully autoclavable
  • Compatible with most standard pipette tips
  • 4 sizes available
Micropette plus features single-channel adjustable volume. Adjustable in various increments depending on size.
Certifications: The MicroPette Plus is calibrated in accordance with ISO 8655, and each pipette includes its own individual test certificate
Accessories information: Pipette Stand is available to accommodate up to 6 pipettes (01-2306)
Specifications
  • Volume:
    100 - 1000 µl
  • Description:
    MicroPette Plus Pipette
  • Increments:
    5 µl