Squibb Dropping Funnels
Socket Size 29/32, 1000 ml
Catalog # 470314-010
Supplier: Wards
: CH0476D
Jump to:
Product Details & Documents
Made of borosilicate glass, with interchangeable plastic stopper and glass stopcock.
Specifications
- Capacity:1000 ml
- Description:Socket Size 29/32