Order Entry
Canada
Orders LinkContactUsLinkComponent
Cryogenic Vials

Cryogenic Vials

1.80 ml, 13×45.7 mm, Yes
Catalog # 470313-434
Supplier:   GSC International
Supplier #:  CRYTB-1.8
Cryogenic Vials
Catalog # 470313-434
Supplier:  GSC International
Cryogenic Vials
Cryogenic Vials

1.80 ml, 13×45.7 mm, Yes

Price
$185.35
Log in for availability
Pack of 500 Case of 2000
Product Details & Documents

Cryogenic vials are used for storing biological samples at extreme temperatures.

  • Leak-proof
  • Printed graduations
  • White area for labeling

Our vials can withstand temperatures as low as –196 °C (approximately –321 °F) and have leak-proof silicone rubber lined caps, printed graduations, and a white area for labeling their contents.

1.8 ml vials measure 45.7 mm (1.8") tall and 13 mm (0.51") in diameter, and our 5 ml vials measure 56.9 mm (2.24") tall and 20 mm (0.79") in diameter.

Specifications
  • Cap included:
    Yes
  • Capacity:
    1.80 ml
  • Diameter ×H:
    13×45.7 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.