Polypropylene Buchner Funnels
125 mm
Catalog # 470313-418
Supplier: GSC International
Supplier #: BFP125
Jump to:
Product Details & Documents
The polypropylene Buchner funnels are two-piece, light-weight, and easy-to-clean.
They are fully autoclavable. The top portion snaps to the bottom forming a vacuum tight seal.
Specifications
- Top diameter:125 mm