Orders LinkContactUsLinkComponent
Objectives for REBEL Microscopes, ECHO
Objectives for REBEL Microscopes, ECHO
20× Plan Apochromat, CSC 0.17, 0.6 mm, N.A. 0.75
Catalog # 76307-582
Supplier:   DISCOVER ECHO INC.
Supplier #:  RMAG9202
Objectives for REBEL Microscopes, ECHO
Catalog # 76307-582
Supplier:  DISCOVER ECHO INC.
Objectives for REBEL Microscopes, ECHO
20× Plan Apochromat, CSC 0.17, 0.6 mm, N.A. 0.75
Price
Inquire For Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Specifications
  • Description:
    20× Plan Apochromat, CSC 0.17
  • Aperture:
    N.A. 0.75
  • Working distance:
    0.6 mm