Orders LinkContactUsLinkComponent
Objectives for REBEL Microscopes, ECHO
Objectives for REBEL Microscopes, ECHO
2.1 mm, N.A. 0.40
Catalog # 76307-580
Objectives for REBEL Microscopes, ECHO
Catalog # 76307-580
Supplier:  
Objectives for REBEL Microscopes, ECHO
2.1 mm, N.A. 0.40
Discontinued
Product Details & Documents
Specifications
  • Aperture:
    N.A. 0.40
  • Working distance:
    2.1 mm