Objectives for REBEL Microscopes, ECHO
20× Plan Achromat Phase, LWD, 7.9 mm, N.A. 0.40
Catalog # 76307-572
Supplier: DISCOVER ECHO INC.
Supplier #: RMAG0213
Jump to:
Product Details & Documents
Specifications
- Description:20× Plan Achromat Phase, LWD
- Aperture:N.A. 0.40
- Working distance:7.9 mm