Orders LinkContactUsLinkComponent
Objectives for REBEL Microscopes, ECHO
Objectives for REBEL Microscopes, ECHO
100× Plan Achromat Oil Phase, CSC 0.17, 0.2 mm, N.A. 1.25
Catalog # 76307-570
Objectives for REBEL Microscopes, ECHO
Catalog # 76307-570
Supplier:  
Objectives for REBEL Microscopes, ECHO
100× Plan Achromat Oil Phase, CSC 0.17, 0.2 mm, N.A. 1.25
Discontinued
Product Details & Documents
Specifications
  • Description:
    100× Plan Achromat Oil Phase, CSC 0.17
  • Aperture:
    N.A. 1.25
  • Working distance:
    0.2 mm