Objectives for REBEL Microscopes, ECHO
100× Plan Achromat Oil, CSC 0.17, 0.2 mm, N.A. 1.25
Catalog # 76307-560
Supplier: DISCOVER ECHO INC.
: RMAG1002
Jump to:
Product Details & Documents
Specifications
- Description:100× Plan Achromat Oil, CSC 0.17
- Aperture:N.A. 1.25
- Working distance:0.2 mm