Trypsinizing Flasks, Bellco Glass
Trypsinizing flask, 50 ml, 50×80 mm
Catalog # 76305-352
Marketsource
Supplier: Bellco Glass
Supplier #: 1992-00050
Jump to:
Product Details & Documents
Manufactured from Type 1 Class A Borosilicate Glass.
- Designed with deep side baffles for optimal agitation
- Large bottom surface for easy access with Bellco large orifice pipette
Specifications
- Description:Trypsinizing flask, 50 ml
- O.D.×H:50×80 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



