Ward's Benchtop Counter
9-Key Benchtop Counter, 32×7.6×5.6 cm
Catalog # 470311-186
Supplier: Heathrow Scientific
Supplier #: HS5360BW
Jump to:
Product Details & Documents
Manual counting made simple.
- Each key records up to 999
- Six and nine key options available
Ward's mechanical, benchtop counters are perfect for blood cells (label included), bacterial colonies, or other repetitive counting. Total window keeps track of the total number of strokes made on all other keys. A bell rings when total reaches 100 Blood cell key labels. 6-Key: baso., eosin., mono., lymph., poly., and total; 9-Key: baso., eosin., myelo., juven., stab., seg., lymph., mono., and total.
Specifications
- Description:9-Key Benchtop Counter
- Dimensions:32×7.6×5.6 cm