Crazy Circuits Classroom Sets: Circuits 101
Product 470348-944 is a replacement for 470311-062 but varies by Brand, Packaging or Quantity.
Product Details & Documents
A complete classroom solution to the daunting task of teaching electronics.
- As with all Crazy Circuits kits, our classroom sets require no soldering
- Parts are connected in a brick-based building environment via our special maker tape conductive tape
- The only additional tools you'll need are scissors
The Circuits 101 Classroom line was designed as a complete classroom solution for teaching electronics. These modular kits are robust sets of materials supported by online lesson plans, projects, activities, diagrams, and videos. Circuits 101 sets each come with one Teacher box and your choice of 4, 9, or 24 Student boxes filled with electronics parts and supplies. The Teacher box includes a full set of Student supplies as well as additional materials. These modular kits are supported by equally robust, NGSS-aligned, online teaching resources designed to facilitate easy classroom use. In addition to lesson plans, the online resources also include activity pages, assessments, diagrams, and videos for classroom use.
Ordering information: Scissors are required but not included.
Delivery information: Each Student box includes 27 Parts + organizer, (5) Coin cell batteries (CR2032), (4) Resistors, (4) Jumbo LEDs, (3) Switches (pushbutton, slide), (2) Transistors (PNP, NPN), (1) Bundle of 5 alligator clips, (1) Photoresistor, (1) Vibrating motor, (1) Roll ⅛” Maker Tape™, (1) Brick baseplate, (1) Coin cell battery holder, (1) Screw terminal, (1) Female header, (1) Potentiometer, Instruction cards/online curricula. Each Teacher box includes Complete contents of Student Box, Large organizer (replaces organizer from Student box), (4) rolls of ⅛” Nylon conductive tape, (20) additional CR2032 batteries.
Specifications
- Description:Crazy Circuits: Circuits 101, 4 pack
- Includes:4 student boxes