Order Entry
Canada
Orders LinkContactUsLinkComponent
Polypropylene Stirring Rods

Polypropylene Stirring Rods

7 mm, 200 mm
Catalog # 470310-638
Supplier:   Wards
Supplier #:  CH0757C
Polypropylene Stirring Rods
Catalog # 470310-638
Supplier:  Wards
Polypropylene Stirring Rods
Polypropylene Stirring Rods

7 mm, 200 mm

Price
$17.95
Log in for availability
Pack of 12
Product Details & Documents

Perfect for mixing liquids and chemicals in the laboratory.

Specifications
  • Ø:
    7 mm
  • Length:
    200 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.