Polypropylene Stirring Rods
10 mm, 250 mm
Catalog # 470310-634
Supplier: Wards
Supplier #: CH0757A
Jump to:
Product Details & Documents
Perfect for mixing liquids and chemicals in the laboratory.
Specifications
- Ø:10 mm
- Length:250 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



