Order Entry
Canada
Orders LinkContactUsLinkComponent
Acetone ACS

Acetone ACS

Glass bottle, 16 L
Catalog # RCRSOA00104X4C
Health
Irritant
Flammable
   Safer Human and Environmental Health
Acetone ACS
Catalog # RCRSOA00104X4C
Supplier:  
   Safer Human and Environmental Health
CAS Number:  67-64-1
Acetone ACS
Acetone ACS

Glass bottle, 16 L

CAS Number:  67-64-1
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BDH ACETONE ACS GRADE GLASS BTL 4L Catalog #BDH1101-4LG

Product BDH1101-4LG is similar to RCRSOA00104X4C but may have a different manufacturer and/or packaging.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Boiling Pt:56.05°C (1013.25 hPa)
  • Melting Pt:–94.7°C
  • Density:0.7880
  • Flash Pt:–20°C
  • UN:1090
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Assay:
    min 99.5 %
  • Color (APHA):
    max 10
  • Solubility in Water:
    To Pass Test
  • Isopropyl Alcohol:
    max 0.05 %
  • Methanol (Methyl Alcohol):
    max 0.05 %
  • Water (H₂O):
    max 0.5 %
  • Aldehyde (as HCHO):
    max 0.002 %
  • Residue after evaporation (Non-Volatile Matter):
    max 0.001 %

Specifications

  • Pack type:
    Glass bottle
  • Size:
    16 L
  • Cat. No.:
    RCRSOA00104X4C
  • Environmental certification:
    Chem21 Solvent selection guide recommended after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.