Acetone ACS
Plastic bottle, 4 L
Supplier: Ricca Chemical
Supplier #: RSOA0010-4A
Acetone ACS
Catalog # RCRSOA0010-4A
Supplier: Ricca Chemical
CAS Number: 67-64-1
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Boiling Pt:56.05°C (1013.25 hPa)
- Melting Pt:–94.7°C
- Density:0.7880
- Flash Pt:–20°C
- UN:1090
- ADR:3,II
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Assay:min 99.5 %
- Color (APHA):max 10
- Solubility in Water:To Pass Test
- Isopropyl Alcohol:max 0.05 %
- Methanol (Methyl Alcohol):max 0.05 %
- Water (H₂O):max 0.5 %
- Aldehyde (as HCHO):max 0.002 %
- Residue after evaporation (Non-Volatile Matter):max 0.001 %
Specifications
- Pack type:Plastic bottle
- Size:4 L
- Cat. No.:RCRSOA0010-4A
- Environmental certification:Chem21 Solvent selection guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



