Order Entry
Canada
Orders LinkContactUsLinkComponent
Storage Bins

Storage Bins

ESD stacking bins, Black, PP, Case of 60, 6.50×4.13×2.95"
Catalog # 10486-477
Supplier:   Treston
Supplier #:  1015-4ESD
Storage Bins
Catalog # 10486-477
Supplier:  Treston
Storage Bins
Storage Bins

ESD stacking bins, Black, PP, Case of 60, 6.50×4.13×2.95"

Price
$887.32
Log in for availability
Case of 60
Minimum order quantity is 1
Product Details & Documents

These high quality, durable Treston bins have been designed for daily use in stores, production facilities and warehouses.

  • Straight sides ensure an efficient use of space
  • Multiple color options make finding the right item easy
  • Open front provides easy access
  • Corrugated base helps retrieve contents, especially smaller sized and flat objects

 : Labels and dividers sold separately, (translucent plastic), labels and blanking piece FP-6M to produce flat surface for adhesive barcode labels on inside rear wall of bins. Contact Avantor representative for more information. Installation is required. Assembly videos available.

Specifications
  • Color:
    Black
  • Description:
    ESD stacking bins
  • Dimensions:
    6.50×4.13×2.95"
  • Material:
    PP
  • Size:
    Case of 60