Order Entry
Canada
Orders LinkContactUsLinkComponent
Storage Bins | Avantor

Storage Bins

Shelf bins, Clear, Polystyrene, Case of 15, 23.62×7.32×3.23"
Catalog # 10117-875
  
Supplier:   Treston
Fournisseur #:  6020-1
Storage Bins
Catalog # 10117-875
Supplier:  Treston
  
Storage Bins
Storage Bins

Shelf bins, Clear, Polystyrene, Case of 15, 23.62×7.32×3.23"

Price
$719.71
Log in for availability
Case of 15
La quantité minimale de commande est de 1
Product Details & Documents

These high quality, durable Treston bins have been designed for daily use in stores, production facilities and warehouses.

  • Straight sides ensure an efficient use of space
  • Multiple color options make finding the right item easy
  • Open front provides easy access
  • Corrugated base helps retrieve contents, especially smaller sized and flat objects

Ordering information: Labels and dividers sold separately, (translucent plastic), labels and blanking piece FP-6M to produce flat surface for adhesive barcode labels on inside rear wall of bins. Contact Avantor representative for more information. Installation is required. Assembly videos available.

Specifications
  • Color:
    Clear
  • Description:
    Shelf bins
  • Dimensions:
    23.62×7.32×3.23"
  • Material:
    Polystyrene
  • Size:
    Case of 15
  • Cat. No.:
    10117-875
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.