Order Entry
Canada
Orders LinkContactUsLinkComponent
Storage Bins

Storage Bins

Shelf bins, Grey, PP, Case of 15, 15.75×7.32×3.23"
Catalog # 10117-797
Supplier:   Treston
Supplier #:  4020-3
Storage Bins
Catalog # 10117-797
Supplier:  Treston
Storage Bins
Storage Bins

Shelf bins, Grey, PP, Case of 15, 15.75×7.32×3.23"

Price
$494.17
Log in for availability
Case of 15
Minimum order quantity is 1
Product Details & Documents

These high quality, durable Treston bins have been designed for daily use in stores, production facilities and warehouses.

  • Straight sides ensure an efficient use of space
  • Multiple color options make finding the right item easy
  • Open front provides easy access
  • Corrugated base helps retrieve contents, especially smaller sized and flat objects

Ordering information: Labels and dividers sold separately, (translucent plastic), labels and blanking piece FP-6M to produce flat surface for adhesive barcode labels on inside rear wall of bins. Contact Avantor representative for more information. Installation is required. Assembly videos available.

Specifications
  • Color:
    Grey
  • Description:
    Shelf bins
  • Dimensions:
    15.75×7.32×3.23"
  • Material:
    PP
  • Size:
    Case of 15
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.