Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Storage Bins

Shelf bins, Red, PP, Case of 15, 11.81×7.32×3.23"
Catalog # 10117-791
Supplier:   Treston
 :  3020-5
Storage Bins
Catalog # 10117-791
Supplier:  Treston
Storage Bins
Storage Bins

Shelf bins, Red, PP, Case of 15, 11.81×7.32×3.23"

Price
$480.81
Log in for availability
Case of 15
 
Product Details & Documents

These high quality, durable Treston bins have been designed for daily use in stores, production facilities and warehouses.

  • Straight sides ensure an efficient use of space
  • Multiple color options make finding the right item easy
  • Open front provides easy access
  • Corrugated base helps retrieve contents, especially smaller sized and flat objects

 : Labels and dividers sold separately, (translucent plastic), labels and blanking piece FP-6M to produce flat surface for adhesive barcode labels on inside rear wall of bins. Contact Avantor representative for more information. Installation is required. Assembly videos available.

Specifications
  • Color:
    Red
  • Description:
    Shelf bins
  • Dimensions:
    11.81×7.32×3.23"
  • Material:
    PP
  • Size:
    Case of 15