Objectives for REVOLVE Microscopes, Echo
4× POL U-Plan fluorite objective, 17 mm, 0.13 NA
Catalog # 76299-622
: JMAG0046
Jump to:
Discontinued
Product Details & Documents
Specifications
- Magnification:4×
- Description:4× POL U-Plan fluorite objective
- Aperture:0.13 NA
- Working distance:17 mm