Objectives for REVOLVE Microscopes, Echo
10× Plan achromat objective, 10.6 mm, 0.25 NA
Catalog # 76299-616
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG0108
Jump to:
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:10×
- Description:10× Plan achromat objective
- Aperture:0.25 NA
- Working distance:10.6 mm