Order Entry
Canada
Orders LinkContactUsLinkComponent
TrueBlack® WB Blocking Buffer Kit, Biotium

TrueBlack® WB Blocking Buffer Kit, Biotium

TrueBlack® WB Blocking Buffer Kit, 10 membranes
Catalog # 76294-168
Supplier:   Biotium
Supplier #:  23013-T
TrueBlack® WB Blocking Buffer Kit, Biotium
Catalog # 76294-168
Supplier:  Biotium
CAS Number:  
TrueBlack® WB Blocking Buffer Kit, Biotium

TrueBlack® WB Blocking Buffer Kit, 10 membranes

CAS Number:  
Price
$192.10
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The TrueBlack® WB blocking buffer kit is a ready-to-use buffer system that provides optimal specificity, sensitivity, and background signal suppression for fluorescence-based western blotting (WB).

  • Blocks non-specific protein binding, reducing background fluorescence over the entire membrane
  • Suppresses background caused by charged fluorescent dyes (performs better than BSA, gelatin, or casein)
  • Reduces antibody cross-reactivity, eliminating non-specific bands
  • Compatible with both polyvinylidene difluoride (PVDF) and nitrocellulose membranes
  • Can be used with antibodies labeled with all types of fluorophores spanning the visible and NIR spectrum
  • Contains non-mammalian blocking agents, for broad secondary antibody compatibility
  • TBS based buffer suitable for phosphoprotein detection

The buffers are designed to achieve optimal specificity and sensitivity by blocking background from the non-specific interaction between the fluorophore-labeled antibodies and the membrane. Non-specific signal in WB can arise from multiple sources, including antibody cross-reactivity with off-target proteins, non-specific antibody adsorption to the membrane, and membrane autofluorescence. Another potential cause of background that is not well-known is the effect of fluorescent dyes themselves on the specificity of labeled antibodies.

Next-generation fluorescent dyes like Alexa Fluor® or CF® dyes often carry multiple negative charges to improve dye solubility and brightness of conjugates. The extra charge carried by the dye can result in non-specific antibody binding and background fluorescence. The TrueBlack® WB Blocking Buffer Kit blocks background from multiple sources including charged dye conjugates. It is especially advantageous for phosphoprotein detection, significantly improving specificity compared to conventional blocking buffers.

Specifications

Specifications

  • Description:
    TrueBlack® WB Blocking Buffer Kit, 10 membranes

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.