VWR® Microfiber String Mop
Product 89496-572 is similar to 76292-772 but manufacturer and specifications may vary.
Product 89214-778 is similar to 76292-772 but manufacturer and specifications may vary.
Product Details & Documents
This Microfiber String Mop is an absorbent and hard wearing mop for cleaning and liquid pick-up in controlled and critical environments.
- Highly absorbent
- Durable
The unique ribbing of the microfiber strands produces a mop that is both low linting and absorbent. The strands are comprised of split fiber polyester filaments, 30 times smaller than cotton fibers. These microfibers work to pick up tiny particles where cotton/rayon blends and polyesters may often roll over the dirt. The solid constructions of the mop, combined with the durable microfiber, allow the mop to be used with strong disinfectants, laundered and re-used.
Documents
Certificates
Search by Lot NumberVWR Cleaning Essentials for Controlled and Critical Environments
Product Literature
Specifications
- Description:Microfiber string mop
- Weight:12 oz.