Order Entry
Canada
Orders LinkContactUsLinkComponent
Polypropylene Strap Assembly, Justrite®

Polypropylene Strap Assembly, Justrite®

Replacement part for gas cylinder equipment, with steel buckle, 1", 54", 2", 1 lb.
Catalog # 76292-440
Supplier:   Justrite
Supplier #:  35406
Polypropylene Strap Assembly, Justrite®
Catalog # 76292-440
Supplier:  Justrite
Polypropylene Strap Assembly, Justrite®
Polypropylene Strap Assembly, Justrite®

Replacement part for gas cylinder equipment, with steel buckle, 1", 54", 2", 1 lb.

Price
$25.19
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

Polypropylene strap assembly provides safety when moving and storing of gas cylinders.

  • Available with either a steel cinch buckle or a nylon side latch buckle

For quick securement of gas cylinders, these heavy-duty polypropylene support straps, 38×1372 mm (1.5×54") are rated at 516 kg (1138 lbs.).

To be used in conjunction with gas cylinder support stands, brackets, hand trucks, carts, and other securement products.

Specifications
  • Description:
    Replacement part for gas cylinder equipment, with steel buckle
  • Height:
    1"
  • Depth:
    54"
  • Weight:
    1 lb.
  • Width:
    2"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.